Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-isobutyrylglycine | 15926-18-8 | MFCD09043666 | 95.0% | 145.16 g/mol | C6H11NO3 | 5 G
SDP

Catalog No. 5000282981
Encompass_Preferred

N-Isobutyrylglycine is an N-acylglycine derivative provided as a research reagent for laboratory and analytical use. It is intended for biochemical and metabolic studies and is supplied with supporting quality documents for traceability. Not for human or clinical use.

  • N-acylglycine derivative.
  • Research use only; not for human or clinical use.
  • CAS number 15926-18-8.
  • Molecular formula C6H11NO3; molecular weight 145.16 g/mol.
  • Supplied with data sheet, certificate of analysis, and safety data sheet.
  • Purity 95.0%.
  • Available in multiple package sizes, including 5 G.

Catalog No. 50-002-82981 Supplier Medchemexpress LLC Supplier No. HYW1419285G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Isobutyrylglycine is an N-acylglycine derivative provided as a research reagent for laboratory and analytical use. It is intended for biochemical and metabolic studies and is supplied with supporting quality documents for traceability. Not for human or clinical use.

  • N-acylglycine derivative.
  • Research use only; not for human or clinical use.
  • CAS number 15926-18-8.
  • Molecular formula C6H11NO3; molecular weight 145.16 g/mol.
  • Supplied with data sheet, certificate of analysis, and safety data sheet.
  • Purity 95.0%.
  • Available in multiple package sizes, including 5 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.