Learn More
Medchemexpress LLC N-Isobutyrylguanosine | 64350-24-9 | 99.7% | 353.33 | 500 G
SDP
Catalog No. 5000404669
Encompass_Preferred
iBu-rG (N-Isobutyrylguanosine) is a nucleoside compound that can be used to synthesize oligoribonucleotides. It is for research use only.
  • Can be used to synthesize oligoribonucleotides.
  • High purity (99.74%).
  • Provided with data sheet, COA, SDS, and handling instructions.
  • Available in both solid and pre-dissolved solution forms.
Catalog No. 50-004-04669 Supplier Medchemexpress LLC Supplier No. HY43057500G
iBu-rG (N-Isobutyrylguanosine) is a nucleoside compound that can be used to synthesize oligoribonucleotides. It is for research use only.
  • Can be used to synthesize oligoribonucleotides.
  • High purity (99.74%).
  • Provided with data sheet, COA, SDS, and handling instructions.
  • Available in both solid and pre-dissolved solution forms.