Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N'-isonicotinoylisonicotinohydrazide | 4329-75-3 | MFCD00534270 | 242.23 g/mol | C12H10N4O2 | 250 MG
SDP

Catalog No. 5000285280
Encompass_Preferred

N'-isonicotinoylisonicotinohydrazide is a small-molecule hydrazide used in research related to reactive oxygen species and antibacterial activity. It appears as a powder and is characterized by a molecular formula of C12H10N4O2 and a molecular weight of 242.23 g/mol. The compound has specific storage recommendations for both powder and solution forms.

  • Molecular formula: C12H10N4O2
  • Molecular weight: 242.23 g/mol
  • Physical form: powder
  • CAS number: 4329-75-3
  • Recommended storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month
  • Research uses: reactive oxygen species and antibacterial studies; relevant pathways include immunology/inflammation and NF-κB

Catalog No. 50-002-85280 Supplier Medchemexpress LLC Supplier No. HYW354203250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N'-isonicotinoylisonicotinohydrazide is a small-molecule hydrazide used in research related to reactive oxygen species and antibacterial activity. It appears as a powder and is characterized by a molecular formula of C12H10N4O2 and a molecular weight of 242.23 g/mol. The compound has specific storage recommendations for both powder and solution forms.

  • Molecular formula: C12H10N4O2
  • Molecular weight: 242.23 g/mol
  • Physical form: powder
  • CAS number: 4329-75-3
  • Recommended storage: powder -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month
  • Research uses: reactive oxygen species and antibacterial studies; relevant pathways include immunology/inflammation and NF-κB

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.