Learn More
ALADDIN SCIENTIFIC CORPORATION N-Isopropylacrylamide, 25g
Catalog No. 5000082517
Encompass_Preferred
N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA pNIPAAm pNIPAA or pNIPAm) thermosensitive polymer- or copolymer-based hydrogels Polymers that contain NIPAM shrink dramatically above 33 C Monomer used in the preparation of thermally sensitive water-swellable hydrogels
Catalog No. 50-000-82517 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. I10681825G
N-Isopropylacrylamide is used to produce poly(N-isopropylacrylamide) (pNIPA pNIPAAm pNIPAA or pNIPAm) thermosensitive polymer- or copolymer-based hydrogels Polymers that contain NIPAM shrink dramatically above 33 C Monomer used in the preparation of thermally sensitive water-swellable hydrogels