Learn More
Medchemexpress LLC N-Isopropylacrylamide-d7 | 1219803-32-3 | 98.1% | 120.20 | 1 MG
SDP
Catalog No. 5000452919
Encompass_Preferred
N-Isopropylacrylamide-d7 is the deuterium labeled N-Isopropylacrylamide. It is intended for research use only and can be utilized as a tracer or as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS. Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis.
  • Applicable in NMR, GC-MS, or LC-MS.
  • Deuterium substitution may impact pharmacokinetic and metabolic profiles.
Catalog No. 50-004-52919 Supplier Medchemexpress LLC Supplier No. HYW010543S1MG
N-Isopropylacrylamide-d7 is the deuterium labeled N-Isopropylacrylamide. It is intended for research use only and can be utilized as a tracer or as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS. Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis.
  • Applicable in NMR, GC-MS, or LC-MS.
  • Deuterium substitution may impact pharmacokinetic and metabolic profiles.