Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Isopropylethylenediamine 98.0+%, TCI America™
SDP

Catalog No. I038110ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I038110ML 10 mL
I038125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I038110ML Supplier TCI America Supplier No. I038110ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19522-67-9
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.181
MDL Number MFCD00008164
InChI Key KDRUIMNNZBMLJR-UHFFFAOYSA-N
Synonym n-isopropylethylenediamine, 2-isopropylaminoethylamine, isopropylaminoethylamine, 1,2-ethanediamine, n-1-methylethyl, ethylenediamine, n-isopropyl, n-isopropylethane-1,2-diamine, 2-aminoethyl propan-2-yl amine, 1,2-ethanediamine, n1-1-methylethyl, 2-aminoethyl isopropyl amine, n1-isopropylethane-1,2-diamine
PubChem CID 88098
ChEBI CHEBI:84288
IUPAC Name N'-propan-2-ylethane-1,2-diamine
SMILES CC(C)NCCN

Specifications

Color Yellow
Boiling Point 138°C
Quantity 10 mL
UN Number 2733
Formula Weight 102.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Isopropylethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.