Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Isopropylmethylamine

Catalog No. 35937850ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-937-850ML 50 mL
35-937-810ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-937-850ML Supplier Sigma Aldrich Supplier No. 35937850ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-isopropylmethylamine is an unsymmetrical amine
  • Pd/c-catalyzed oxidative cross double carbonylation of n-isopropylmethylamine with alcohols has been reported.n-isopropylmethylamine is suitable for use in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (raft) polymerization
  • It is suitable for use in the synthesis of novel 5-ethoxy-n,n-dialkyl-[α,α,α,α-h4]-tryptamines and their [α,α,α,α,αd4]-counterparts following the speeter and anthony procedure

Specifications

CAS 4747-21-1
Density 0.702 g/mL (at 25°C (literature))
Boiling Point 50°C to 53°C (lit.)
Molecular Formula C4H11N
Refractive Index n20/D 1.384 (literature)
Linear Formula (CH3)2 CHNHCH3
MDL Number MFCD00042859
Quantity 50 mL
Synonym N-Methylisopropylamine
Molecular Weight (g/mol) 73.14
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.