Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Isopropylmethylamine, 98%

Catalog No. AC439650100
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439650100 10 mL Glass bottle
AC439650500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC439650100 Supplier Thermo Scientific Chemicals Supplier No. 439650100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 4747-21-1 | C4H11N | 73.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4747-21-1
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
InChI Key XHFGWHUWQXTGAT-UHFFFAOYSA-N
Synonym n-isopropylmethylamine, n-methylisopropylamine, methylisopropylamine, 2-propanamine, n-methyl, isopropylmethylamine, 2-methylaminopropane, methyl propan-2-yl amine, ethylamine, n,1-dimethyl, unii-j3ol90426g, n-methylisopropyylamine
PubChem CID 78485
IUPAC Name N-methylpropan-2-amine
SMILES CC(C)NC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.7020g/mL
Boiling Point 50°C to 53°C
Flash Point -32°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.3830 to 1.3850 (20°C, 589nm)
Linear Formula CH3NHCH(CH3)2
Quantity 10 mL
Specific Gravity 0.702
Formula Weight 73.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Isopropylmethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
EINECSNumber 225-266-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.