Learn More
Medchemexpress LLC HY-W015464 100mg Medchemexpress, N-Isovaleroylglycine CAS:16284-60-9 Purity:>98%
SDP
Catalog No. 501934602
Encompass_Preferred
Medchemexpress, HY-W015464 100mg N-Isovaleroylglycine CAS:16284-60-9 N-Isovaleroylglycine is an acyl glycine and could be used as a biomarker for the predispositon for weight gain and obesity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4602 Supplier Medchemexpress LLC Supplier No. HYW015464100MG
Medchemexpress, HY-W015464 100mg N-Isovaleroylglycine CAS:16284-60-9 N-Isovaleroylglycine is an acyl glycine and could be used as a biomarker for the predispositon for weight gain and obesity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.