Learn More
Medchemexpress LLC N-(ketocaproyl)-D,L-homoserine lactone | 76924-95-3 | MFCD03788849 | 97.0% | 213.24 g·mol⁻¹ | C10H15NO4 | 5 MG
SDP
Catalog No. 5000262184
Encompass_Preferred
N-(Ketocaproyl)-DL-homoserine lactone is a homoserine lactone quorum-sensing autoinducer and a ligand of LuxR-family receptors. Supplied as a solid for research use, it is employed in microbiology and signaling assays to study bacterial communication and receptor binding. Typical identifiers include CAS 76924-95-3, formula C10H15NO4, and molecular weight ≈213.24 g·mol⁻¹.
  • Used to probe quorum sensing pathways in bacteria.
  • Suitable for receptor binding and signaling assays.
  • Supplied as a white to off-white solid for convenient handling.
  • Stable when stored frozen at -20 °C to preserve integrity.
  • Available in small research quantities for assay development.
  • High purity as reported by suppliers; verify certificate of analysis before critical use.
Catalog No. 50-002-62184 Supplier Medchemexpress LLC Supplier No. HY1294055MG
N-(Ketocaproyl)-DL-homoserine lactone is a homoserine lactone quorum-sensing autoinducer and a ligand of LuxR-family receptors. Supplied as a solid for research use, it is employed in microbiology and signaling assays to study bacterial communication and receptor binding. Typical identifiers include CAS 76924-95-3, formula C10H15NO4, and molecular weight ≈213.24 g·mol⁻¹.
  • Used to probe quorum sensing pathways in bacteria.
  • Suitable for receptor binding and signaling assays.
  • Supplied as a white to off-white solid for convenient handling.
  • Stable when stored frozen at -20 °C to preserve integrity.
  • Available in small research quantities for assay development.
  • High purity as reported by suppliers; verify certificate of analysis before critical use.