Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(-Ketocaproyl)-DL-homoserine lactone, MilliporeSigma™ Supelco™

Catalog No. 111015477 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5477 50 mg
11-101-5478 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5477 Supplier MilliporeSigma Supelco Supplier No. K325550MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 76924-95-3
Physical Form Neat
Linear Formula C10H15 NO4
Technique Gas Chromatography
MDL Number MFCD03788849
Synonym N-(3-Oxohexanoyl)-DL-homoserine lactone
Type Analytical Standard
Solvent or Matrix Neat
Industry Type Drug Analysis
Molecular Weight (g/mol) 213.23
Quantity 50 mg
Formula Weight 213.23
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.