Learn More
Cayman Chemical N ketocaproylLHomoserine lac
SDP
Catalog No. 502888842
Encompass_Preferred
A bacterial quorum-sensing signaling molecule; produced in the marine bioluminescence bacterium V. fischeri
Catalog No. 50-288-8842 Supplier Cayman Chemical Supplier No. 1001120725 MG
A bacterial quorum-sensing signaling molecule; produced in the marine bioluminescence bacterium V. fischeri