Learn More
Medchemexpress LLC N-(L-3-trans-propylcarbamoyl-oxirane-2-carbonyl)-L-isoleucyl-L-proline | 134448-10-5 | MFCD00797531 | 99.9% | 383.44 g·mol⁻¹ | C18H29N3O6 | 25 MG
SDP
Catalog No. 5000245189
Encompass_Preferred
CA-074 is a potent, selective inhibitor of cathepsin B (Ki 2-5 nM) supplied for research use as a powder or DMSO solution. It is intended for in vitro enzyme inhibition studies and as a control compound in cell-based assays; follow recommended storage to maintain activity.
  • Potent cathepsin B inhibition (Ki 2-5 nM).
  • High purity for research applications.
  • Available as powder or DMSO solution for flexible handling.
  • Stable when stored under recommended frozen conditions.
  • Suitable as a control in biochemical and cellular assays.
Catalog No. 50-002-45189 Supplier Medchemexpress LLC Supplier No. HY10335025MG
CA-074 is a potent, selective inhibitor of cathepsin B (Ki 2-5 nM) supplied for research use as a powder or DMSO solution. It is intended for in vitro enzyme inhibition studies and as a control compound in cell-based assays; follow recommended storage to maintain activity.
  • Potent cathepsin B inhibition (Ki 2-5 nM).
  • High purity for research applications.
  • Available as powder or DMSO solution for flexible handling.
  • Stable when stored under recommended frozen conditions.
  • Suitable as a control in biochemical and cellular assays.