Learn More
Medchemexpress LLC N-lactoyl-phenylalanine-13C6 | 183241-73-8 | 98.5% | 243.21 | 5 MG
SDP
Catalog No. 5000361054
Encompass_Preferred
N-Lactoyl-Phenylalanine-13C6 is a 13C labeled N-Lactoyl-Phenylalanine. It is a blood-derived signaling metabolite that can be induced by exercise, and can reduce obesity and improve glucose tolerance.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-61054 Supplier Medchemexpress LLC Supplier No. HY150012S15MG
N-Lactoyl-Phenylalanine-13C6 is a 13C labeled N-Lactoyl-Phenylalanine. It is a blood-derived signaling metabolite that can be induced by exercise, and can reduce obesity and improve glucose tolerance.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.