Learn More
Medchemexpress LLC N-Lactoyl-Phenylalanine (Lac-Phe) | 183241-73-8 | 99.9% | 237.25 | 1 ML
SDP
Catalog No. 5000359457
Encompass_Preferred
N-Lactoyl-Phenylalanine (Lac-Phe) is an endogenous signaling metabolite found in blood that is induced by exercise. It has been shown to reduce obesity and improve glucose tolerance.
  • Blood-derived signaling metabolite.
  • Induced by exercise.
  • Reduces obesity.
  • Improves glucose tolerance.
  • Specifically suppresses energy intake in obese mice without affecting energy expenditure pathways (at 50 mg/kg, i.p., once a day for 10 days).
  • Results in reduced cumulative food intake and body weight in diet-induced obese mice.
  • Exhibits improved glucose homeostasis and reduced adiposity in treated mice.
  • For research use only.
Catalog No. 50-003-59457 Supplier Medchemexpress LLC Supplier No. HY15001210MM/1ML
N-Lactoyl-Phenylalanine (Lac-Phe) is an endogenous signaling metabolite found in blood that is induced by exercise. It has been shown to reduce obesity and improve glucose tolerance.
  • Blood-derived signaling metabolite.
  • Induced by exercise.
  • Reduces obesity.
  • Improves glucose tolerance.
  • Specifically suppresses energy intake in obese mice without affecting energy expenditure pathways (at 50 mg/kg, i.p., once a day for 10 days).
  • Results in reduced cumulative food intake and body weight in diet-induced obese mice.
  • Exhibits improved glucose homeostasis and reduced adiposity in treated mice.
  • For research use only.