Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-lauroyl-D-erythro-sphingosine | 74713-60-3 | 99.8% | 481.79 | C30H59NO3 | 5 MG
SDP

Catalog No. 5000212697
Encompass_Preferred

C12-Ceramide (N-lauroyl-D-erythro-sphingosine) is a short-chain ceramide used as a research-grade sphingolipid standard and biochemical reagent. Supplied as a purified solid, it is intended for use in studies of ceramide signaling, sphingolipid metabolism, and related cellular and biochemical assays.

  • High purity suitable for analytical and biological applications.
  • Molecular weight 481.79 g/mol.
  • Molecular formula C30H59NO3.
  • Available in small research pack sizes, including 5 mg.
  • Suitable as a reference standard and reagent in sphingolipid research.

Catalog No. 50-002-12697 Supplier Medchemexpress LLC Supplier No. HY1003535MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C12-Ceramide (N-lauroyl-D-erythro-sphingosine) is a short-chain ceramide used as a research-grade sphingolipid standard and biochemical reagent. Supplied as a purified solid, it is intended for use in studies of ceramide signaling, sphingolipid metabolism, and related cellular and biochemical assays.

  • High purity suitable for analytical and biological applications.
  • Molecular weight 481.79 g/mol.
  • Molecular formula C30H59NO3.
  • Available in small research pack sizes, including 5 mg.
  • Suitable as a reference standard and reagent in sphingolipid research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.