Learn More
Medchemexpress LLC N-Lauroyl-D-erythro-sphingosine | 74713-60-3 | 99.8% | 481.79 g·mol⁻¹ | C30H59NO3 | 10 MG
SDP
Catalog No. 5000212696
Encompass_Preferred
C12-Ceramide (N-Lauroyl-D-erythro-sphingosine) is a naturally occurring ceramide used as an analytical standard and a bioactive lipid reagent for cell-based studies. It is applied in research on ceramide-mediated signaling, can enhance doxorubicin cytotoxicity in cancer cell lines, and is used in studies of lipid metabolism disorders.
  • High purity analytical standard for research assays.
  • Well-characterized molecular properties for reproducibility.
  • Soluble in DMSO at 2.5 mg/mL with ultrasonic assistance.
  • Stable as a powder at -20°C for long-term storage.
  • White to off-white solid for straightforward handling.
Catalog No. 50-002-12696 Supplier Medchemexpress LLC Supplier No. HY10035310MG
C12-Ceramide (N-Lauroyl-D-erythro-sphingosine) is a naturally occurring ceramide used as an analytical standard and a bioactive lipid reagent for cell-based studies. It is applied in research on ceramide-mediated signaling, can enhance doxorubicin cytotoxicity in cancer cell lines, and is used in studies of lipid metabolism disorders.
  • High purity analytical standard for research assays.
  • Well-characterized molecular properties for reproducibility.
  • Soluble in DMSO at 2.5 mg/mL with ultrasonic assistance.
  • Stable as a powder at -20°C for long-term storage.
  • White to off-white solid for straightforward handling.