Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-lauroylglycine (N-dodecanoyl-glycine) | 7596-88-5 | MFCD00153470 | 98.0% | 257.37 g·mol⁻¹ | C14H27NO3 | 250 MG
SDP

Catalog No. 5000282892
Encompass_Preferred

N-Lauroylglycine (N-dodecanoyl-glycine) is an N-acylglycine derivative supplied as a high-purity research reagent. It is provided as a solid suitable for biochemical, analytical, and metabolomic studies. For research use only; not for human or clinical use.

  • Purity 98.0%.
  • Chemical formula C14H27NO3.
  • Molecular weight 257.37 g·mol⁻¹.
  • CAS number 7596-88-5.
  • Available in small-scale quantities (250 mg, 500 mg, 1 g, and larger).
  • Datasheet, COA, and SDS available.

Catalog No. 50-002-82892 Supplier Medchemexpress LLC Supplier No. HYW141880250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Lauroylglycine (N-dodecanoyl-glycine) is an N-acylglycine derivative supplied as a high-purity research reagent. It is provided as a solid suitable for biochemical, analytical, and metabolomic studies. For research use only; not for human or clinical use.

  • Purity 98.0%.
  • Chemical formula C14H27NO3.
  • Molecular weight 257.37 g·mol⁻¹.
  • CAS number 7596-88-5.
  • Available in small-scale quantities (250 mg, 500 mg, 1 g, and larger).
  • Datasheet, COA, and SDS available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.