Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI Chemicals N-Lauroylsarcosine, Reagent, 90.0% Purity, 500 g

Catalog No. 502781215
Encompass_Preferred

N-Lauroylsarcosine is a reagent-grade chemical compound typically used for laboratory research applications. This moisture-sensitive material is supplied in a clear glass bottle and requires storage under inert gas at temperatures between 0°C and 10°C.

  • Formula: C15H29NO3
  • Purity: ≥ 90.0% (GC)
  • Molecular weight: 271.40 g/mol
  • Physical state: Solid
  • Melting point: 46°C
  • Solubility: Shows almost transparency in methanol
  • Hazardous: Classified as acute toxicity category 2

Catalog No. 50-278-1215 Supplier Grainger Supplier No. 19KN74
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Lauroylsarcosine is a reagent-grade chemical compound typically used for laboratory research applications. This moisture-sensitive material is supplied in a clear glass bottle and requires storage under inert gas at temperatures between 0°C and 10°C.

  • Formula: C15H29NO3
  • Purity: ≥ 90.0% (GC)
  • Molecular weight: 271.40 g/mol
  • Physical state: Solid
  • Melting point: 46°C
  • Solubility: Shows almost transparency in methanol
  • Hazardous: Classified as acute toxicity category 2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.