Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Lauroylsarcosine, 97% (Ultra Pure Grade), Sodium Salt, MP Biomedicals

Catalog No. ICN19400950 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19400950 50 g
ICN19400980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19400950 Supplier MP Biomedicals, Inc Supplier No. 0219400950
Add to Cart
Add to Cart

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15H28NNaO3
Molecular Weight (g/mol) 293.38
MDL Number MFCD00042728
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
Synonym Sarkosyl NL-97, Sodium N-lauroyl sarcosinate
PubChem CID 23668817
IUPAC Name sodium 2-(N-methyldodecanamido)acetate
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 97%
Formula Weight 293.4
Grade Ultra Pure
Chemical Name or Material N-Lauroylsarcosine, sodium salt
Recommended Storage Store at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.