Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Lauroylsarcosine, MP Biomedicals™

Catalog No. ICN19400990 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19400990 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19400990 Supplier MP Biomedicals, Inc Supplier No. 0219400990
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anionic detergent used for the solubilization and separation of membrane proteins and glycoproteins.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15H28NNaO3
Molecular Weight (g/mol) 293.38
MDL Number MFCD00042728
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
Synonym sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
PubChem CID 23668817
IUPAC Name sodium 2-(N-methyldodecanamido)acetate
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

Quantity 500 g
Percent Purity ≥97%
Grade Ultra Pure
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.