Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Lauroylsarcosine Sodium Salt

Catalog No. 6174550G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-745-50G 50 g
61-745-250G 250 g
61-745-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 61-745-50G Supplier Sigma Aldrich Supplier No. 6174550G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Lauroylsarcosine is an anionic surfactant with an ability to denature proteins. Due to its microbicidal property, N-lauroylsarcosine is being considered as a potent anti-microbicide in topical formulations, especially against sexually transmitted diseases (STDs).N-Lauroylsarcosine sodium salt has been used to study the complexing ability of a novel amphiphilic +a-cyclodextrin with anionic surfactants.Anionic surfactant.

Specifications

CAS 137-16-6
Molecular Formula C15H28NNaO3
Linear Formula CH3(CH2)10CON(CH3)CH2COONa
MDL Number MFCD00042728
Quantity 50 g
Synonym N-Dodecanoyl-N-methylglycine sodium salt; Sarkosyl NL
Molecular Weight (g/mol) 293.38 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.