Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-lauryldiethanolamine | 1541-67-9 | MFCD00059002 | >95.0% | 273.45 g/mol | C16H35NO2 | 100 G
SDP

Catalog No. 5000282090
Encompass_Preferred

N-Lauryldiethanolamine is an aliphatic tertiary amine used as an antistatic agent and surfactant. Supplier listings report bioactivity including inhibitory effects on cell growth and lysosomotropic behavior; consult safety data sheets for handling and toxicity information.

  • Antistatic agent and surfactant
  • Aminodiol structure; molecular formula C16H35NO2
  • Molecular weight 273.45 g/mol
  • CAS number 1541-67-9
  • Typically supplied in 100 G packages
  • Common purity: >95.0% (GC)

Catalog No. 50-002-82090 Supplier Medchemexpress LLC Supplier No. HYW335972100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Lauryldiethanolamine is an aliphatic tertiary amine used as an antistatic agent and surfactant. Supplier listings report bioactivity including inhibitory effects on cell growth and lysosomotropic behavior; consult safety data sheets for handling and toxicity information.

  • Antistatic agent and surfactant
  • Aminodiol structure; molecular formula C16H35NO2
  • Molecular weight 273.45 g/mol
  • CAS number 1541-67-9
  • Typically supplied in 100 G packages
  • Common purity: >95.0% (GC)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.