Learn More
Medchemexpress LLC N-lauryldiethanolamine | 1541-67-9 | MFCD00059002 | 98.0% | 273.45 g·mol-1 | C16H35NO2 | 500 G
SDP
Catalog No. 5000280792
Encompass_Preferred
N-Lauryldiethanolamine is a colorless to light yellow liquid used as an antistatic agent and laboratory reagent. It exhibits lysosomotropic behavior and has reported inhibitory effects on cell growth at micromolar concentrations; it is provided at research-grade purity for formulation and cellular studies.
  • Antistatic and surfactant properties suitable for formulation studies.
  • Reported lysosomotropic behavior useful for cellular uptake research.
  • High reported purity (98.0%) for research-grade applications.
  • Liquid form facilitates handling and incorporation into formulations.
  • CAS-registered chemical for unambiguous identification and compliance.
  • Available in multiple pack sizes to support small- and bulk-scale work.
Catalog No. 50-002-80792 Supplier Medchemexpress LLC Supplier No. HYW335972500G
N-Lauryldiethanolamine is a colorless to light yellow liquid used as an antistatic agent and laboratory reagent. It exhibits lysosomotropic behavior and has reported inhibitory effects on cell growth at micromolar concentrations; it is provided at research-grade purity for formulation and cellular studies.
  • Antistatic and surfactant properties suitable for formulation studies.
  • Reported lysosomotropic behavior useful for cellular uptake research.
  • High reported purity (98.0%) for research-grade applications.
  • Liquid form facilitates handling and incorporation into formulations.
  • CAS-registered chemical for unambiguous identification and compliance.
  • Available in multiple pack sizes to support small- and bulk-scale work.