Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-lauryldiethanolamine | 1541-67-9 | MFCD00059002 | 98.0% | 273.45 g/mol | C16H35NO2 | 250 G
SDP

Catalog No. 5000280578
Encompass_Preferred

N-Lauryldiethanolamine is an antistatic agent and surfactant-grade reagent used in laboratory research. It exhibits lysosomotropic behavior and has reported inhibitory effects on cell growth at micromolar concentrations; follow the SDS for handling and safety information.

  • High purity: 98.0%.
  • Molecular weight: 273.45 g/mol.
  • Molecular formula: C16H35NO2.
  • Antistatic and surfactant properties useful for formulation and dispersion.
  • Shows lysosomotropic behavior and cellular growth inhibition at micromolar levels.
  • Available in laboratory-scale pack sizes, including 250 g.

Catalog No. 50-002-80578 Supplier Medchemexpress LLC Supplier No. HYW335972250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Lauryldiethanolamine is an antistatic agent and surfactant-grade reagent used in laboratory research. It exhibits lysosomotropic behavior and has reported inhibitory effects on cell growth at micromolar concentrations; follow the SDS for handling and safety information.

  • High purity: 98.0%.
  • Molecular weight: 273.45 g/mol.
  • Molecular formula: C16H35NO2.
  • Antistatic and surfactant properties useful for formulation and dispersion.
  • Shows lysosomotropic behavior and cellular growth inhibition at micromolar levels.
  • Available in laboratory-scale pack sizes, including 250 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.