Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Lauryldiethanolamine 95.0+%, TCI America™
SDP

Catalog No. L002025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L002025G 25 g
L00205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L002025G Supplier TCI America Supplier No. L002025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1541-67-9
Molecular Formula C16H34NO2
Molecular Weight (g/mol) 272.45
MDL Number MFCD00059002
InChI Key ASGZLXKBMOBPBV-UHFFFAOYSA-N
PubChem CID 352309
IUPAC Name dodecylidenebis(2-hydroxyethyl)azanium
SMILES CCCCCCCCCCCC=[N+](CCO)CCO

Specifications

Boiling Point 195°C
Quantity 25 g
Synonym N,N-Bis(2-hydroxyethyl)dodecylamine, N-Dodecyldiethanolamine, 2,2′-(Dodecylazanediyl)diethanol
Formula Weight 273.46
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Lauryldiethanolamine
EINECSNumber (2)-0353&(7)-0060
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.