Learn More
Cayman Chemical N-Lithocholyl-L-Leucine, Purity: ≥95%, CAS: 2095154-73-5, 10 mg
SDP
Catalog No. 502945863
Encompass_Preferred
A bile acid conjugate; has been used in the formation of hydrogels
Catalog No. 50-294-5863 Supplier Cayman Chemical Supplier No. 4093110 MG
A bile acid conjugate; has been used in the formation of hydrogels