Learn More
Medchemexpress LLC N-Mal-N-bis(PEG2-NHS ester) | 2182601-73-4 | 98.9% | C29H38N4O15 | 100 MG
SDP
Catalog No. 5000353092
Encompass_Preferred
N-Mal-N-bis(PEG2-NHS ester) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for a target protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-53092 Supplier Medchemexpress LLC Supplier No. HY140571100MG
N-Mal-N-bis(PEG2-NHS ester) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for a target protein. They function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only