Learn More
Medchemexpress LLC N-Mal-N-bis(PEG4-NH-Boc) | 2128735-27-1 | 97.0% | C37H66N4O15 | 25 MG
SDP
Catalog No. 5000352656
Encompass_Preferred
N-Mal-N-bis(PEG4-NH-Boc) is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system. PROTACs achieve this by using two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to the target protein.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Contains two ligands connected by a linker for E3 ubiquitin ligase and target protein binding
  • For research use only
Catalog No. 50-003-52656 Supplier Medchemexpress LLC Supplier No. 25MGHY140575
N-Mal-N-bis(PEG4-NH-Boc) is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are molecules designed to selectively degrade target proteins by leveraging the intracellular ubiquitin-proteasome system. PROTACs achieve this by using two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to the target protein.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Contains two ligands connected by a linker for E3 ubiquitin ligase and target protein binding
  • For research use only