Learn More
Medchemexpress LLC N-Mal-N-bis(PEG4-NHS ester) | 2112738-60-8 | ≥95.0% | C37H54N4O19 | 100 MG
SDP
Catalog No. 5000353049
Encompass_Preferred
N-Mal-N-bis(PEG4-NHS ester) is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It is designed for research applications, leveraging the intracellular ubiquitin-proteasome system for targeted protein degradation.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Connects ligands for E3 ubiquitin ligase and target proteins
  • Employs ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-53049 Supplier Medchemexpress LLC Supplier No. HY140572100MG
N-Mal-N-bis(PEG4-NHS ester) is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It is designed for research applications, leveraging the intracellular ubiquitin-proteasome system for targeted protein degradation.
  • PEG-based PROTAC linker
  • Used for PROTAC synthesis
  • Connects ligands for E3 ubiquitin ligase and target proteins
  • Employs ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only