Learn More
AdipoGen N-Me bisCF3sulfonylimide
SDP
Supplier:  AdipoGen CDXM0154G025
Encompass_Preferred
Chemical. CAS 537031-78-0. Formula C3H3F6NO4S2. MW 295.18. Synthetic Powerful alkylation agent. Used for direct methylation and trifluoroethylation of imidazole and pyridine derivatives to produce a variety of room temperature ionic liquids RTILs.
Catalog No. 50-256-0489