Learn More
Medchemexpress LLC N-Me-L-Ala-maytansinol | 77668-69-0 | MFCD00864836 | 99.9% | 650.16 | C32H44ClN3O9 | 5 MG
SDP
Catalog No. 5000202340
Encompass_Preferred
N-Me-L-Ala-maytansinol is a hydrophobic, cell-permeable maytansinoid used as a cytotoxic payload for preparing antibody-drug conjugates (ADCs) in research applications. It is supplied as a high-purity solid and as a DMSO solution for laboratory use.
  • Hydrophobic, cell-permeable maytansinoid payload.
  • High purity suitable for conjugation chemistry.
  • Available as solid (multiple mg sizes) and as a 10 mM solution in DMSO.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-002-02340 Supplier Medchemexpress LLC Supplier No. HY1266635MG
N-Me-L-Ala-maytansinol is a hydrophobic, cell-permeable maytansinoid used as a cytotoxic payload for preparing antibody-drug conjugates (ADCs) in research applications. It is supplied as a high-purity solid and as a DMSO solution for laboratory use.
  • Hydrophobic, cell-permeable maytansinoid payload.
  • High purity suitable for conjugation chemistry.
  • Available as solid (multiple mg sizes) and as a 10 mM solution in DMSO.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for clinical or human use.