Learn More
Medchemexpress LLC N-Me-N-bis-PEG4 | 2055046-22-3 | C17H37NO8 | 250 MG
SDP
Catalog No. 5000352830
Encompass_Preferred
N-Me-N-bis-PEG4 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds designed with two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, while the other targets the protein of interest. These compounds leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52830 Supplier Medchemexpress LLC Supplier No. 250MGHY140578
N-Me-N-bis-PEG4 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds designed with two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, while the other targets the protein of interest. These compounds leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins