Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-me-thalidomide 4-fluoride | 2244520-92-9 | MFCD34563580 | 99.4% | 290.25 | C14H11FN2O4 | 250 MG
SDP

Catalog No. 5000281731
Encompass_Preferred

N-Me-Thalidomide 4-fluoride is a thalidomide-based cereblon (E3 ligase) ligand used in PROTAC design and in the synthesis of related research compounds. Supplied as a characterized solid for research use, the compound is accompanied by analytical data and safety documentation to support laboratory workflows.

  • High purity (99.4%) suitable for research applications.
  • Molecular formula C14H11FN2O4 and molecular weight 290.25 Da.
  • Available in multiple pack sizes from 250 mg to 10 g.
  • Analytical documentation: COA, HNMR, and RP-HPLC provided.
  • Safety documentation: SDS and handling instructions available.
  • Designed for use as a cereblon ligand in PROTAC conjugation chemistry.

Catalog No. 50-002-81731 Supplier Medchemexpress LLC Supplier No. HYW460193250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Me-Thalidomide 4-fluoride is a thalidomide-based cereblon (E3 ligase) ligand used in PROTAC design and in the synthesis of related research compounds. Supplied as a characterized solid for research use, the compound is accompanied by analytical data and safety documentation to support laboratory workflows.

  • High purity (99.4%) suitable for research applications.
  • Molecular formula C14H11FN2O4 and molecular weight 290.25 Da.
  • Available in multiple pack sizes from 250 mg to 10 g.
  • Analytical documentation: COA, HNMR, and RP-HPLC provided.
  • Safety documentation: SDS and handling instructions available.
  • Designed for use as a cereblon ligand in PROTAC conjugation chemistry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.