Learn More
Medchemexpress LLC N-Me-thalidomide 4-fluoride | 2244520-92-9 | MFCD34563580 | 99.4% | 290.25 g/mol | C14H11FN2O4 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW4601931G
Encompass_Preferred
N-Me-Thalidomide 4-fluoride is a research-grade small molecule ligand for E3 ligase used in chemical biology and PROTAC development. It functions as a building block in the synthesis of targeted degraders and related small-molecule tools. Supplied for laboratory research use only.
  • Ligand for E3 ligase.
  • Useful building block for synthesis of targeted degraders and anti-inflammatory agents.
  • High reported purity (99.41%).
  • Molecular formula C14H11FN2O4; molecular weight 290.25 g/mol.
  • Available in research pack sizes suitable for small-scale synthesis.
Catalog No. 50-002-81734