Learn More
Medchemexpress LLC N-Me-Thalidomide 4-fluoride | 2244520-92-9 | MFCD34563580 | 99.4% | 290.25 g/mol | C14H11FN2O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW460193500MG
Encompass_Preferred
N-Me-Thalidomide 4-fluoride is a thalidomide-derived ligand for E3 ligase (Cereblon) used as a building block in targeted protein degradation chemistry and small-molecule conjugation. It is supplied as a research-grade small molecule with characterization data for medicinal chemistry and chemical biology applications.
  • Ligand for E3 ligase (Cereblon) for PROTAC synthesis.
  • High reported purity suitable for research use.
  • Molecular formula C14H11FN2O4 and molecular weight 290.25 g/mol.
  • Available in multiple pack sizes, including 250 MG, 500 MG, and 1 G.
  • Supplied with associated safety and batch documentation.
Catalog No. 50-002-84130