Learn More
Sigma Aldrich N-Methoxy-N,2-dimethyl-oxazole-4-carboxamide
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 1,3-dichloropropene (1,3-d) is a clear, light-straw colored liquid
- It is a soil fumigant useful for preplanting control of parasitic nematodes
- Co-formulations of 1,3-d and chloropicrin are widely employed for preplant fumigation in the production of high-value crops.1,3-dichloropropene may be employed in commercial fumigant preparations
- It is used as pesticide in a study to investigate the photocatalytic degradation of water-soluble pesticides (cymoxanil, methomyl, oxamyl, dimethoate, pyrimethanil and telone) at pilot-plant scale
Specifications
Specifications
| CAS | 875553-59-6 |
| Molecular Formula | C7H10N2O3 |
| Linear Formula | C7H10N2O3 |
| MDL Number | MFCD11040624 |
| Quantity | 250 mL |
| Molecular Weight (g/mol) | 170.17 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.