Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methoxycarbonylmaleimide 98.0+%, TCI America™
SDP

Catalog No. M23331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23331G 1 g
M23335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23331G Supplier TCI America Supplier No. M23331G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55750-48-6
Molecular Formula C6H5NO4
Molecular Weight (g/mol) 155.109
MDL Number MFCD00042755
InChI Key LLAZQXZGAVBLRX-UHFFFAOYSA-N
PubChem CID 580610
IUPAC Name methyl 2,5-dioxopyrrole-1-carboxylate
SMILES COC(=O)N1C(=O)C=CC1=O

Specifications

Melting Point 66°C
Quantity 1 g
Formula Weight 155.11
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Methoxycarbonylmaleimide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.