Learn More
Medchemexpress LLC N-methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin | 72252-90-5 | MFCD00077135 | 97.0% | C31H41N5O9 | 10MG
SDP
Catalog No. 5000260971
Encompass_Preferred
MeOSuc-AAPV-AMC is a fluorogenic peptide substrate for measuring elastase activity. Cleavage by elastase releases 7-amino-4-methylcoumarin (AMC), producing a sensitive fluorescent signal (Ex 380 nm, Em 460 nm) suitable for in vitro enzyme assays and cell-based studies.
  • Fluorogenic readout using AMC (Ex 380 nm, Em 460 nm).
  • Sensitive detection of human leukocyte and porcine pancreatic elastase.
  • Km approximately 362 μM for elastase, suitable for kinetic assays.
  • Supplied as a powder with high purity and good solubility in DMSO (100 mg/mL).
  • Recommended storage sealed, protected from moisture and light at -20 °C to -80 °C depending on duration.
  • Convenient small-quantity packaging for assay development and optimization.
Catalog No. 50-002-60971 Supplier Medchemexpress LLC Supplier No. HY13686510MG
MeOSuc-AAPV-AMC is a fluorogenic peptide substrate for measuring elastase activity. Cleavage by elastase releases 7-amino-4-methylcoumarin (AMC), producing a sensitive fluorescent signal (Ex 380 nm, Em 460 nm) suitable for in vitro enzyme assays and cell-based studies.
  • Fluorogenic readout using AMC (Ex 380 nm, Em 460 nm).
  • Sensitive detection of human leukocyte and porcine pancreatic elastase.
  • Km approximately 362 μM for elastase, suitable for kinetic assays.
  • Supplied as a powder with high purity and good solubility in DMSO (100 mg/mL).
  • Recommended storage sealed, protected from moisture and light at -20 °C to -80 °C depending on duration.
  • Convenient small-quantity packaging for assay development and optimization.