Learn More
Medchemexpress LLC N-methyl-2-(pyridin-2-yl)-N-(2-(pyridin-2-yl)ethyl)ethanamine | 5452-87-9 | MFCD28157290 | 98.4% | 241.33 g/mol | C15H19N3 | 100 MG
SDP
Catalog No. 5000240800
Encompass_Preferred
Betahistine EP Impurity C is an analytical reference standard used for identification and quantification of a known impurity related to betahistine. The compound has molecular formula C15H19N3, molecular weight 241.33 g/mol, and a batch-reported purity of 98.38%. It is intended for method development, calibration, and impurity profiling in chromatographic and mass spectrometric assays.
  • Analytical reference standard for impurity identification and quantification.
  • Suitable for HPLC, GC, and MS method development.
  • High reported purity (98.38%) for accurate calibrations.
  • Defined molecular weight and formula to support analytical characterization.
  • Provided in small-quantity packaging for laboratory use.
Catalog No. 50-002-40800 Supplier Medchemexpress LLC Supplier No. HY107495100MG
Betahistine EP Impurity C is an analytical reference standard used for identification and quantification of a known impurity related to betahistine. The compound has molecular formula C15H19N3, molecular weight 241.33 g/mol, and a batch-reported purity of 98.38%. It is intended for method development, calibration, and impurity profiling in chromatographic and mass spectrometric assays.
  • Analytical reference standard for impurity identification and quantification.
  • Suitable for HPLC, GC, and MS method development.
  • High reported purity (98.38%) for accurate calibrations.
  • Defined molecular weight and formula to support analytical characterization.
  • Provided in small-quantity packaging for laboratory use.