Learn More
Medchemexpress LLC N-methyl-2-pyridin-2-yl-N-(2-pyridin-2-ylethyl)ethanamine | 5452-87-9 | MFCD28157290 | 98.4% | 241.33 | C15H19N3 | 10 MG
SDP
Catalog No. 5000241430
Encompass_Preferred
Betahistine EP Impurity C is an analytical reference standard representing a characterized impurity of the active pharmaceutical ingredient betahistine. Supplied in small reference quantities for laboratory use, it is intended for analytical method development, identity confirmation, and quality control in pharmaceutical research.
  • High reported purity suitable for analytical applications.
  • Molecular formula C15H19N3 and molecular weight 241.33 for precise identification.
  • Available in milligram-scale quantities for standard preparation.
  • Supplied with batch-specific certificate of analysis for traceability.
Catalog No. 50-002-41430 Supplier Medchemexpress LLC Supplier No. HY10749510MG
Betahistine EP Impurity C is an analytical reference standard representing a characterized impurity of the active pharmaceutical ingredient betahistine. Supplied in small reference quantities for laboratory use, it is intended for analytical method development, identity confirmation, and quality control in pharmaceutical research.
  • High reported purity suitable for analytical applications.
  • Molecular formula C15H19N3 and molecular weight 241.33 for precise identification.
  • Available in milligram-scale quantities for standard preparation.
  • Supplied with batch-specific certificate of analysis for traceability.