Learn More
Sigma Aldrich Fine Chemicals Biosciences N-Methyl-2-pyridone-5-carboxamide | MFCD00120693 | 50 MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SMB0134550MG
Encompass
N-Methyl-2-pyridone-5-carboxamide | Purity: Research grade | Mol Wt: 136.15 g/mol | CAS No: 2670-11-7 | MDL No: MFCD00120693 | Quantity: 50mg
Catalog No. NC2594754