Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Methyl-2-pyrrolecarboxaldehyde

Catalog No. 1300525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-005-25G 5 g
13-005-225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-005-25G Supplier Sigma Aldrich Supplier No. 1300525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Methyl-2-pyrrolecarboxaldehyde was used to prepare 3-(1-methyl-1H-pyrrol-2-yl)propanoic acid.

Specifications

CAS 1192-58-1
Density 1.016 g/mL (at 25°C (literature))
Boiling Point 87°C to 90°C (22 mmHg)
Molecular Formula C6H7NO
Linear Formula C6H7NO
MDL Number MFCD00003087
Quantity 5 g
Molecular Weight (g/mol) 109.13
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.