Learn More
Medchemexpress LLC N-methyl-2-pyrrolidone | 872-50-4 | MFCD00003193 | 100.0% | 99.13 g/mol | C5H9NO | 50g
SDP
Catalog No. 5000266784
Encompass_Preferred
N-Methylpyrrolidone (1-Methyl-2-pyrrolidinone) a five-membered cyclic amide is an orally active organic polar solvent with teratogenicity and toxicity N-Methylpyrrolidone is low in acute toxicity with a LD50 value of 3914 mg/kg in rats and of 4050 mg/kg in mice N-Methylpyrrolidone is extensively used in the manufacture of adhesives paints fuels and pharmaceuticals[1][2][3][4]
Catalog No. 50-002-66784 Supplier Medchemexpress LLC Supplier No. HYY127550G
N-Methylpyrrolidone (1-Methyl-2-pyrrolidinone) a five-membered cyclic amide is an orally active organic polar solvent with teratogenicity and toxicity N-Methylpyrrolidone is low in acute toxicity with a LD50 value of 3914 mg/kg in rats and of 4050 mg/kg in mice N-Methylpyrrolidone is extensively used in the manufacture of adhesives paints fuels and pharmaceuticals[1][2][3][4]