Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methyl-3'-hydroxycoclaurine hydrochloride | 138644-99-2 | 351.82 | C18H22ClNO4 | 500 UG
SDP

Catalog No. 5000283986
Encompass_Preferred

N-Methyl-3'-hydroxycoclaurine hydrochloride is the hydrochloride salt of a coclaurine-related small molecule described as an impurity and used in research and synthetic chemistry as a drug intermediate or analytical reference. The compound is provided for laboratory and drug-discovery workflows where related intermediates, impurities, or standards are required.

  • Hydrochloride salt form for handling and formulation consistency.
  • Small-molecule impurity related to coclaurine chemistry.
  • Suitable for use as a drug intermediate or analytical standard.
  • Molecular formula C18H22ClNO4 and molecular weight 351.82.
  • CAS number 138644-99-2 for unambiguous identification.
  • Certificate of analysis provides storage and batch-specific purity information.

Catalog No. 50-002-83986 Supplier Medchemexpress LLC Supplier No. HYW587979500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Methyl-3'-hydroxycoclaurine hydrochloride is the hydrochloride salt of a coclaurine-related small molecule described as an impurity and used in research and synthetic chemistry as a drug intermediate or analytical reference. The compound is provided for laboratory and drug-discovery workflows where related intermediates, impurities, or standards are required.

  • Hydrochloride salt form for handling and formulation consistency.
  • Small-molecule impurity related to coclaurine chemistry.
  • Suitable for use as a drug intermediate or analytical standard.
  • Molecular formula C18H22ClNO4 and molecular weight 351.82.
  • CAS number 138644-99-2 for unambiguous identification.
  • Certificate of analysis provides storage and batch-specific purity information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.