Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methyl-3-(pyrido[2,3-d]pyrimidin-7-yl)benzamide derivative | 1567915-38-1 | 100.0% | 474.55 | C26H30N6O3 | 5MG
SDP

Catalog No. 5000238138
Encompass_Preferred

MTI-31 is a potent, selective inhibitor of mTORC1 and mTORC2 used in preclinical research to probe PI3K/Akt/mTOR signaling and antitumor activity. It is supplied as a solid research compound intended for laboratory in vitro and in vivo studies; consult the manufacturer's datasheet for handling and safety details.

  • Selective mTORC1/mTORC2 inhibitor with reported Kd 0.20 nM.
  • Used to study PI3K/Akt/mTOR pathway and antitumor responses.
  • Molecular formula C26H30N6O3 and molecular weight 474.55.
  • Purity 99.98% (manufacturer reported).
  • Solid, white to yellow; supplied as a 5 mg quantity.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-38138 Supplier Medchemexpress LLC Supplier No. HY1260775MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MTI-31 is a potent, selective inhibitor of mTORC1 and mTORC2 used in preclinical research to probe PI3K/Akt/mTOR signaling and antitumor activity. It is supplied as a solid research compound intended for laboratory in vitro and in vivo studies; consult the manufacturer's datasheet for handling and safety details.

  • Selective mTORC1/mTORC2 inhibitor with reported Kd 0.20 nM.
  • Used to study PI3K/Akt/mTOR pathway and antitumor responses.
  • Molecular formula C26H30N6O3 and molecular weight 474.55.
  • Purity 99.98% (manufacturer reported).
  • Solid, white to yellow; supplied as a 5 mg quantity.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.