Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-4-nitro-o-toluidine 97.0+%, TCI America™
SDP

Catalog No. M09535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M09535G Supplier TCI America Supplier No. M09535G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10439-77-7
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00059083
InChI Key NJYYBTRBCMPYCJ-UHFFFAOYSA-N
Synonym N,2-Dimethyl-4-nitroaniline, 2-(Methylamino)-5-nitrotoluene
PubChem CID 25282
IUPAC Name N,2-dimethyl-4-nitroaniline
SMILES CNC1=CC=C(C=C1C)[N+]([O-])=O

Specifications

Melting Point 138°C
Color Yellow
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-4-nitro-o-toluidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.