Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl[5-(2-furyl)pyrid-2-yl]methylamine 95%, Thermo Scientific™

Catalog No. CC55146DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC55146DA 1 g Amber Glass Bottle
CC55146CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC55146DA Supplier Thermo Scientific Chemicals Supplier No. CC55146DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 892502-04-4
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD10700054
InChI Key DOKOUBYFAJTDEI-UHFFFAOYSA-N
Synonym 5-furan-2-yl pyridin-2-yl methyl methyl amine, 2-pyridinemethanamine,5-2-furanyl-n-methyl, 5-fur-2-yl-2-methyl aminomethyl pyridine, 5-2-furyl 2-pyridyl methyl methylamine, 5-fur-2-yl pyridin-2-yl-n-methylmethylamine, 1-5-furan-2-yl pyridin-2-yl-n-methylmethanamine
PubChem CID 26343611
IUPAC Name 1-[5-(furan-2-yl)pyridin-2-yl]-N-methylmethanamine
SMILES CNCC1=NC=C(C=C1)C2=CC=CO2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 188.23
Percent Purity 90%
Chemical Name or Material n-methyl[5-(2-furyl)pyrid-2-yl]methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.