Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-bis(trifluoroacetamide), ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018471 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8471 5 mL
11-101-8296 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8471 Supplier MilliporeSigma Supelco Supplier No. M07895ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00000412
Synonym MBTFA (N-methyl-bis(trifluoroacetamide)or BTFA (bis(TFA)); MBTFA

Specifications

Density 1.547 g/mL (at 25°C)
CAS 685-27-8
Boiling Point 121°C to 122°C
Flash Point 45°C
Packaging Glass Bottle
Linear Formula CF3 CON(CH3)COCF3
Quantity 5 mL
Formula Weight 223.07
Percent Purity ≥97.0% (GC)
Grade For GC derivatization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.