Learn More
Medchemexpress LLC N-methyl-β-carboline-3-carboxamide | 78538-74-6 | MFCD00055075 | 99.4% | 225.25 g/mol | C13H11N3O | 25 MG
SDP
Catalog No. 5000212570
Encompass_Preferred
FG 7142 is a benzodiazepine inverse agonist used as a research tool to study GABAA receptor pharmacology and neural signaling. It preferentially binds α1-containing GABAA receptors, modulates GABA-induced chloride flux, and affects dopaminergic activity in rodent models. The compound is supplied as a light yellow solid of high purity with recommended storage conditions.
Catalog No. 50-002-12570 Supplier Medchemexpress LLC Supplier No. HY10099125MG
FG 7142 is a benzodiazepine inverse agonist used as a research tool to study GABAA receptor pharmacology and neural signaling. It preferentially binds α1-containing GABAA receptors, modulates GABA-induced chloride flux, and affects dopaminergic activity in rodent models. The compound is supplied as a light yellow solid of high purity with recommended storage conditions.